Gelatin cell culture flasks, Corning® BioCoat®
Supplier: Corning
354488 356488
734-0162EA 97.26 EUR
734-0162 734-0293
Gelatin cell culture flasks, Corning® BioCoat®
Flasks Cell Culture Flasks
PS, coated with porcine gelatin, sterile, non-pyrogenic.
  • Gelatin substrate enhances the attachment of a variety of normal and transfected cell types
  • Pre-treatment with gelatin eliminates time consuming preparation, saving time and money
  • Lot-to-lot consistency ensures reproducible results
Corning® BioCoat™ gelatin provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of normal and transfected cell types, including vascular endothelial, muscle, embryonic stem (ES) and F9More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk