Bulk packaged serological pipettes, Falcon®
Supplier: Corning
357506 357508 357515 357529 357530
734-0335EA 252.99 EUR
734-0335 734-0337 734-0338 734-0344 734-0345
Bulk packaged serological pipettes, Falcon®
Pipettes Serological Pipettes
The pipettes graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.
  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0,1 EU/ml
  • Color-coded package and markings for ease of identification
Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduationsMore Product Information