96-Well T-cell activation plates, Corning® BioCoat™
Supplier: Corning
354725
734-1075EA 565.98 EUR
734-1075
96-Well T-cell activation plates, Corning® BioCoat™
Microplates HTS Microplates
Corning BioCoat T-Cell activation plates are precoated with high quality BD Pharmingen™ CD3 antibodies. Available for use with mouse or human T-cells, Corning BioCoat T-Cell activation plates offer lot-to-lot consistency and come individually packaged with lids for ease of use.
  • Tested for ability to proliferate mouse splenocytes or human PBMCs
  • Tested and found negative for bacteria and fungi
Applications include:
- T-Cell activation
- Cytokine production
- Cytokine mRNA quantitation
- Co-stimulation
- Studies of drug effects on T-cell function