Strips of 8 PCR tubes and cap strips, Corning® Thermowell® GOLD
Supplier: Corning
6542 6547 3740 3741 3742 3743 3748
734-1760EA 598.06 EUR
734-1760 734-1765 732-5578 732-5579 732-5580 732-5581 732-5586
Strips of 8 PCR tubes and cap strips, Corning® Thermowell® GOLD
Tubes PCR Tubes
Tube strips consist of eight 0,2 ml thin wall PP tubes connected together. Dual connectors between adjacent tubes eliminate inadvertent breakage during sample handling. Tube strips are designed for precise fit in thermal cyclers to optimise heat transfer. Thermowell® GOLD cap strips are sold separately. Tube strips are tested and certified to be free of DNase/RNase contamination and are autoclavable to 121 °C.
The optically clear, 8-well flat cap strips feature an ultra-clear surface ideal for real-time PCR and fluorescent and luminescent applications.
Informace pro objednávání: More Product Information