Cell culture dishes, Nunclon™ Sphera™
Supplier: Thermo Fisher Scientific
174943 174944 174945
734-2770EA 223 EUR
734-2770 734-2771 734-2772
Cell culture dishes, Nunclon™ Sphera™
Culture Dishes Cell Culture Dishes
The Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment. The surface supports many different cell types and their ability to generate spheroids.
  • Optimises spheroid formation
  • Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
  • Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimises variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth, providing a better modelMore Product Information