96-well permeable support systems and plates, HTS Transwell®
Supplier: Corning
3374 3380 3381 3384 3385 3386 3387 3388 3391 3392 7369
734-4071EA 201.5 EUR
734-4071 734-4074 734-4075 734-4078 734-4079 734-4080 734-4081 734-4082 734-4083 734-4084 734-3204
96-well permeable support systems and plates, HTS Transwell®
Microplates Cell Culture Plates
Permeable supports for HTS (high throughput screening) applications are excellent for ADMET screening, drug transport, and other assays.
The HTS Transwell® 96-well permeable support features 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit. Available with polyester or polycarbonate membranes in a variety of pore sizes. The 0,143 cm² well membrane area provides significantly more surface area than comparable systems. Large apical and basolateral access ports allow efficient media sampling and facilitateMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk