Microplates, 96-well, Nunc™ Edge 2.0
Supplier: Thermo Fisher Scientific
167425 167542 167554 167574 267427 267544 267556 267576 267566 267578
735-0326EA 160 EUR
735-0326 735-0327 735-0328 735-0329 735-0330 735-0331 735-0332 735-0334 735-0333 735-0335
Microplates, 96-well, Nunc™ Edge 2.0
Microplates Cell Culture Plates
The Nunc™ Edge plate has a design that minimises evaporation and the risks associated with the edge effect. When filled with sterile water or media, the surrounding moat acts as an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
  • Reduce edge effect for full-plate results
  • Yield viable cells with well-to-well