Microplate readers, multi-mode, SpectraMax® iD5
Supplier: Molecular Devices
ID5-STD ID5-BL-INJ ID5BLINJSCACA3 ID5-BL ID5-INJ ID5BLSCACAD3 ID5INJSCACAD3 ID5BLINJSCACAD ID5-BL-SC-ACAD ID5INJSCACAD
735-0358EA 0 EUR
735-0358 735-0378 MLDVID5BLINJSCACA3 735-0377 735-0379 MLDVID5BLSCACAD3 MLDVID5INJSCACAD3 MLDVID5BLINJSCACAD MLDVID5-BL-SC-ACAD MLDVID5INJSCACAD
Microplate readers, multi-mode, SpectraMax® iD5
Microplate Readers
The SpectraMax® iD5 multi-mode microplate reader is a complete laboratory solution to increase your research capabilities and comes with built-in absorbance, fluorescence, luminescence, time-resolved fluorescence (TRF) and tuneable fluorescence polarisation (FP) read modes. In addition, the SpectraMax® iD5 can be expanded to include bottom-read luminescence, TR-FRET, HTRF®, BRET, dual luciferase reporter assays with injectors and Western blot detection.
  • Easily set up protocols, run experiments, view tutorial videos, and control user access with the large
More Product Information
Order Now
SPECIFICATIONS
Model SpectraMax® iD5
Light source Xenon flash lamp
Environmentally Preferable
Measuring modes Abs, FI, Lum, TRF, FP, FRET and TR-FRET (endpoint and kinetic reading); Abs, FI, Lum, TRF (spectral scanning)
Detector PMT and/or photodiode
Wavelength range 230 - 1000 nm
Wavelength selection 1,0 nm increments
Wavelength accuracy ±2,0 nm
Wavelength bandwidth 4,0 nm
Incubation range 5 °C above ambient to 65 °C
Photometric range 0 - 4,0 OD
Plate types 6- to 384-well plates
Shaking Linear, orbital and double orbital
Temperature accuracy ±1 °C @ 37 °C
Temperature uniformity ±0,75 °C
W×D×H 532×598×401 mm
Weight 40 kg