Multiwell cell culture plates, Falcon®
Supplier: Corning
353043 353046 353047 353078 353225 353226 353230 351143 351146 351147 353224 353934 353935 351178
391-0006EA 207.01 EUR
391-0006 734-0019 734-0020 734-0028 734-0055 734-0056 734-0058 734-0947 734-0948 734-0949 734-0054 736-2025 736-2026 734-0956
Multiwell cell culture plates, Falcon®
Microplates Cell Culture Plates
Standard tissue culture (TC) and Corning® Primaria™ surface treatments is available. These plates are provided with various colours to suit various detection methods.
  • Crystal-grade virgin polystyrene
  • Nonpyrogenic
  • Sterilised by gamma irradiation
  • Growth area and volume are nominal
Individual and ready-stack (RS) trays are PET (Code 1) and recyclable.
Balení: Convenient, peel-open medical-style packaging for individual product presentation.