Sodium orthovanadate 99,9 Metals basis

Supplier: Thermo Fisher Scientific
Warning

81104.14 81104.22 81104.36
81104.14EA 105 EUR
81104.14 81104.22 81104.36
Sodium orthovanadate 99,9 Metals basis
Sodium orthovanadate

Purity: 99.9% CAS No.: 13721-39-6 Molecular Formula: Na3O4V Formula Weight: 183.94 Physical Form: Granular Appearance: White Melting Point: 850-866° MDL No.: MFCD00003511


Inhibitor of protein tyrosine phosphatase and alkaline phosphate

Formula: Na₃O₄V
MW: 183.91 g/mol
Melting Pt: 850…866 °C
Storage Temperature: Ambient
MDL Number: MFCD00003511
CAS Number: 13721-39-6
EINECS: 237-287-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR