Tough-Spots® Label, Rolls
Supplier: DIVERSIFIED BIOTECH
T-SPOTS T-SPOTS-L T-SPOTS-R T-SPOTS-G T-SPOTS-Y T-SPOTS-B T-SPOTS-50 T-SPOTS-50R T-SPOTS-50G T-SPOTS-50Y T-SPOTS-50B T-SPOTS-O T-SPOTS-P T-SPOTS-50O T-SPOTS-44 T-SPOTS-S SPRL-2100 SPRL-1100 SPRL-2000 T-SPOTS-C
817-2141EA 75.9 EUR
817-2141 817-0389 817-0392 817-0388 817-0394 817-0386 817-2136 817-2139 817-2138 817-2140 817-2137 817-0390 817-0391 817-0108 817-0385 817-0393 817-0384 817-0382 817-0383 817-0387
Tough-Spots® Label, Rolls
Labels
These Tough-Spots® PVC label rolls are available in a variety of sizes, colors and formats.
  • Temperature resistant from −196 to +80 °C
  • Accept writing from most lab markers
The dispensing box assists in labeling individual tubes used in small experiments.