VWR® Preservation System, Cryoinstant

Supplier: VWR Chemicals

822075ZAEA 174 EUR
822075ZA 822070ZA 822071ZA 822072ZA 822073ZA 822074ZA 820006ZA
VWR® Preservation System, Cryoinstant
Microbiology Test Systems Microorganism Tests

Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in contaminated cultures, loss of viability and, even more importantly, loss of original characteristics by introducing biochemical and genetic changes. Cryoinstant consists of 25 porous beads in a vial filled with a broth containing glycerol. Microorganisms will bind to the porous surface of the beads. The excess broth is aspirated, and the vials stored at temperatures down to –70 °C in a freezer.


  • Quick and easy to use
  • Colour coded
  • Storage down to –70 °C
  • Quality controlled for fertility and sterility


Balení: Cryoinstant is packed in packs of 50 vials of 2 ml.

Order Now

SPECIFICATIONS

Environmentally Preferable
Shelf life 1440 days 1440 1440
Storage temperature 8...25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR