E/E absolute 99,5, TechniSolv®, pure

Supplier: VWR Chemicals
Danger

Synonyms: Ethyl alcohol

83813.360_TEA 54.6 EUR
83813.360_T 83813.410_T 83813.440_T 83813.360 83813.410 83813.440 83813.550 83813.930
E/E absolute 99,5, TechniSolv®, pure
Ethanol

Upozornení: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Formula: H₃CCH₂OH
MW: 46.07 g/mol
Boiling Pt: 78.3 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay (V/V) Min. 99.5 %
Density (20/20) 0.788 - 0.794
Water Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR