Acetonitrile 99,9, HiPerSolv CHROMANORM® HPLC_prep

Supplier: VWR Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

84533.550EA 0 EUR
84533.550 84533.460 84533.950
Acetonitrile 99,9, HiPerSolv CHROMANORM® HPLC_prep
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Limited Time Sale

Select items below are currently on sale

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Acidity Max. 0.0005 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.02 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR