1,4-Dioxane 99,8 Unstabilised, SPECTRONORM® Spectroscopy

Supplier: VWR Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

Filter 0,2 um_Nitrogen_filled

84715.290EA 247 EUR
84715.290
1,4-Dioxane 99,8 Unstabilised, SPECTRONORM® Spectroscopy
1,4-Dioxane

SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1 °C (1013 hPa)
Melting Pt: 11.8 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Residue on evaporation Max. 0.0005 %
Water Max. 0.02 %
Transmittance (270 nm) Min. 70 %
Transmittance (280 nm) Min. 85 %
Transmittance (290 nm) Min. 95 %
Transmittance (300 nm) Min. 97 %
Transmittance (310 nm) Min. 98 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR