Bismuth(III) nitrate basic, AnalaR NORMAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate

84838.180EA 93.9 EUR
84838.180
Bismuth(III) nitrate basic, AnalaR NORMAPUR®
Bismuth (III) nitrate basic
Formula: Bi₅O(OH)₉(NO₃)₄
MW: 1461.99 g/mol
Density: 4.928 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064844
CAS Number: 1304-85-4
EINECS: 215-136-8
UN: 1477
ADR: 5.1 II

Order Now

Specification Test Results

Assay (calculated as Bi) 71.0 - 74.0 %
Identification Passes test
Loss on drying (105°C) Max. 3 %
Substances not precipitated by ammonia Max. 1 %
Cl (Chloride) Max. 0.02 %
Ag (Silver) Max. 25 ppm
Cu (Copper) Max. 50 ppm
Pb (Lead) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR