Acetonitrile with 0,05% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® HPLC
Supplier: VWR Chemicals
Nitrogen_filled
84863.320EA 251 EUR
84863.320
Acetonitrile with 0,05% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® HPLC
Acetonitrile with 0,05% (v/v) trifluoroacetic acid
Order Now
Specification Test Results
Assay Min. 99.9 %
Nominal concentration (TFA; v/v) 0.0475 to 0.0525 %
Evaporation residue Max. 0.0001 %
Water Max. 0.005 %
Transmittance (195 nm) Min.20 %
Transmittance (200 nm) Min. 50 %
Transmittance (245 nm) Min. 80 %
Transmittance (260 nm) Min. 95 %
Transmittance (270 nm) Min. 99 %