VWR®, Fluoride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
84966.180EA 60.3 EUR
84966.180 84966.260
VWR®, Fluoride 1,000 mg/L in Water standard for ion chromatography
Standards Anionic Standards Fluoride Standards
Certifikace: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.