N,N-Dimethylformamide 99,0-100,00, PESTINORM® GC_HS

Supplier: VWR Chemicals
Danger

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

Filter 0,2 um_Nitrogen_filled

85395.290EA 124 EUR
85395.290 85395.320
N,N-Dimethylformamide 99,0-100,00, PESTINORM® GC_HS
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: –60.5 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (DIN 51755) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 99.8%
Identity (IR) Complying
Residue after evaporation max. 0.0003%w/w
Water (KF) max. 0.05%w/w
Color (APHA Pt-Co) max. 10
GC-HS (Each class 1 solvent) max. 1µg/g
GC-HS (Each class 2 solvent) max. 10µg/g
GC-HS (Each class 3 solvent) max. 50µg/g
Application HS-GC (standard measurement) Passes test
Headspace test for O.V.I's Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR