Trisodium citrate concentrated solution, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate

854C-20ML
854C-20MLEA 108 EUR
854C-20ML
Trisodium citrate concentrated solution, Sigma-Aldrich®
tri-Sodium citrate

Citrate Concentrated Solution. pH approx. 3.0 when diluted 1:50. For the preparation of a 60% citrate buffered acetone solution used as a fixative for blood smears.

Formula: C₆H₅Na₃O₇
MW: 258.07 g/mol
Melting Pt: 150
Flash Pt: 290
Storage Temperature: Ambient
MDL Number: MFCD00012462
CAS Number: 68-04-2
Merck Index: 12,08746
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR