VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
85539.1000EA 890 EUR
85539.1000 85540.1000 85541.1000 85542.1000 85543.1000 85544.1000 85525.1000 85526.1000 85527.1000 85528.1000 85529.1000 85530.1000 85531.1000 85532.1000 85533.1000 85534.1000 85535.1000 85536.1000 85537.1000 85538.1000 85535.0100 85536.0100 85537.0100 85538.0100 85539.0100 85540.0100 85541.0100 85542.0100 85543.0100 85544.0100 85525.0100 85526.0100 85527.0100 85528.0100 85529.0100 85530.0100 85531.0100 85532.0100 85533.0100 85534.0100
VWR® Kjeldahl Tablets
Digestion Systems Kjeldahl Tablets and Reagents
Kjeldahl tablets are catalysts that promote the digestion step of the Kjeldahl method for nitrogen determination.
  • Wide range of catalysts available
  • Different sizes of tablet available
  • Tablet format is easy to use
The Kjeldahl method is used in a wide range of analyses including analysis of proteins, nitrogenated organic materials and also, after reduction, for analysis of nitrates and nitrites. Therefore, the method is used widely in applications from food and beverage testing to agricultural and environmental analysis.
Total Kjeldahl Nitrogen (TKN) is often used as a parameter in wastewater and soil analysis.
A wide range of catalysts have been developed over the 130 year history of this method to provide ever more rapid and safe digestion methods for particular sets of digestion conditions. Avantor offers a comprehensive range of 'ready to use' tablets to meet the optimal requirements for the sulphuric acid decomposition phase.