VWR®, Colour reference solutions acc. to Ph. Eur., ready to use
Supplier: VWR Chemicals
85752.001EA 552 EUR
85752.001 85753.001 85754.001 85755.001 85756.001 88364.001
VWR®, Colour reference solutions acc. to Ph. Eur., ready to use
Standards Colour Reference and Spectroscopy Standards
These solutions are developed and produced according to chapter 2.2.2. 'Degree of colouration of liquids' of the European Pharmacopoeia. (brown, brown-yellow, green-yellow, red and yellow). These kits are ready to use, and are dedicated to help you save time and money because preparation of these solutions is very complex.
  • Easy to use kits
  • Ready to use solutions
  • Stable - shelf life of 36 months
  • Certificate of Analysis provided with the kit
The kits are easy to use: Just add the sample into the empty vial provided with the kit and compare the colour with the reference solution in the rack.