VWR® AVS TITRINORM, Cesium chloride/lanthanum chloride buffer solution acc. to schinkel for atomic absorption spectroscopy (aas)
Supplier: VWR Chemicals
85773.290EA 219 EUR
85773.290
VWR® AVS TITRINORM, Cesium chloride/lanthanum chloride buffer solution acc. to schinkel for atomic absorption spectroscopy (aas)
Buffers Ionisation Buffers
This material is intended for use as an ionisation buffer for AAS
Assay Lanthanum (La) – 100 g/l
Assay Cesium (Cs) – 7.9 g/l
Impurities
- Ca (Calcium) < 5.0 mg/l
- Cd (Cadmium) < 0.2 mg/l
- Co (Cobalt) < 1.0 mg/l
- Cr (Chromium) < 1.0 mg/l
- Cu (Copper) < 0.2 mg/l
- Fe (Iron) < 1.0 mg/l
- K (Potassium) < 1.0 mg/l
- Li (Lithium) < 0.2 mg/l
- Mg (Magnesium) < 1.0 mg/l
- Mn (Manganese) < 0.2 mg/l
- Na (Sodium) < 2.0 mg/l
- Ni (Nickel) < 1.0 mg/l
- Sr (Strontium) < 1.0 mg/l
- Zn (Zinc) < 2.0 mg/l