VWR® AVS TITRINORM, Calcium Standard Solution, 1000 mg/L Ca in dil. Nitric Acid (from Ca(NO₃)₂), Standard for AAS
Supplier: VWR Chemicals
86667.260EA 44.8 EUR
86667.260 86667.180
VWR® AVS TITRINORM, Calcium Standard Solution, 1000 mg/L Ca in dil. Nitric Acid (from Ca(NO₃)₂), Standard for AAS
Standards Cationic Standards Calcium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%