VWR®, Tungsten standard solution, 1000 mg/L W in nitric acid with hydrofluoric acid (each max. 1%) AVS TITRINORM standard for AAS
Supplier: VWR Chemicals
86722.260EA 69.8 EUR
86722.260 86722.180
VWR®, Tungsten standard solution, 1000 mg/L W in nitric acid with hydrofluoric acid (each max. 1%) AVS TITRINORM standard for AAS
Standards Cationic Standards Tungsten Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%