HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Hexamethyldisilazane

87078.150EA 28.9 EUR
87078.150 87078.230 87078.290
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%, GPR RECTAPUR®
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 126
Melting Pt: -82
Density: 0.78 g/cm³ (20 °C)
Flash Pt: 6
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 1992
ADR: 3 (6.1, 8) II
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98
Density (20/4) 0.774 - 0.775

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR