Benzyl chloride 99, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: (Chloromethyl)benzene, Chlorophenylmethane, α-Chlorotoluene

87103.290EA 44.6 EUR
87103.290 87103.180 87103.320 87103.460
Benzyl chloride 99, GPR RECTAPUR®
Benzyl chloride
Formula: (C₆H₅)CH₂Cl
MW: 126.59 g/mol
Boiling Pt: 179.3
Melting Pt: -41.2
Flash Pt: 52
Storage Temperature: Ambient
MDL Number: MFCD00000889
CAS Number: 100-44-7
UN: 1738
ADR: 6.1 (8) II
Merck Index: 13,01131

Order Now

Specification Test Results

Assay (GC, area%) ≥ 99%
Appearance: Clear colourless to pale yellow liquid Passes test
Density (20 °C) 1,097 - 1,101
Acidity ≤ 0,002 meq/g
Non-volatile matter ≤ 0,05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR