Cobalt(II) oxalate dihydrate, Reagent Grade

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Cobalt(II) oxalate dihydrate

87758.06 87758.14 87758.22 87758.36
87758.06EA 33.9 EUR
87758.06 87758.14 87758.22 87758.36
Cobalt(II) oxalate dihydrate, Reagent Grade
Cobalt (II) oxalate dihydrate

CAS No.: 5965-38-8 Molecular Formula: CoC2O4.2H2O Formula Weight: 182.98 (146.95anhy) Physical Form: Powder Density: 3.021 Odour: Odourless Merck Reference: 14,2445 UN No.: 3288 MDL No.: MFCD00050793


Stabilizer for HCN, temperature indicator

Formula: C₂CoO₄·2H₂O
MW: 182.98 g/mol (146.95 anhy)
Melting Pt: 229…231 °C (dec.)
Density: 3.021
Storage Temperature: Ambient
MDL Number: MFCD03414148
CAS Number: 5965-38-8
EINECS: 212-409-3
UN: 3288
ADR: 6.1 III
Merck Index: 14,2445

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR