Zinc chloride 97,0, granules, AR® ACS, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
Danger
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
8780-20EA 0 EUR
8780-20 8780-02 8780-04
Zinc chloride 97,0, granules, AR® ACS, Macron Fine Chemicals™
Zinc chloride
Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 13,10185
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 97.0 %
ACS - Assay (ZnCl₂) ≥97.0 %
Ammonium (NH₄) ≤0.005 %
Calcium (Ca) ≤0.06 %
Insoluble Matter ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.005 %
Magnesium (Mg) ≤0.01 %
Nitrate (NO₃) ≤0.003 %
Oxychloride Passes Test
Potassium (K) ≤0.02 %
Sodium (Na) ≤0.05 %
Solubility Passes Test
Sulfate (SO₄) ≤0.01 %
Appearance (off-white to gray granules)