Iodinated potassium iodide solution (mixture Potassium iodide : Iodine) (40:20) g/l aqueous solution Reag. Ph. Eur. 1070503
Supplier: VWR Chemicals
87910.180EA 137 EUR
87910.180
Iodinated potassium iodide solution (mixture Potassium iodide : Iodine) (40:20) g/l aqueous solution Reag. Ph. Eur. 1070503
iodinated Potassium iodide solution (mixture of 60g/l Potassium iodide and 20 g/l Iodine in aqueous solution)
Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.