Cadmium standard solution, 1000 mg/L Cd in 5% nitric acid (from Cd), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific

88056.AE 88056.AP
88056.AEEA 45.4 EUR
88056.AE 88056.AP
Cadmium standard solution, 1000 mg/L Cd in 5% nitric acid (from Cd), standard for AAS, Specpure®
Standards Cationic Standards Cadmium Standards

Molecular Formula: Cd in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010914


Certifikace: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00010914

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR