Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific

88112.AE 88112.AP
88112.AEEA 53.8 EUR
88112.AE 88112.AP
Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for AAS, Specpure®
Standards Cationic Standards Tin Standards

Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00133862


Certifikace: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00133862

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR