VWR®, TOC standard solutions
Supplier: VWR Chemicals
88222.180EA 85.5 EUR
88222.180 88222.260 88223.180 88223.260 88224.180 88224.260 88225.180 88225.260 88226.180 88226.260 88227.180 88227.260 88228.180 88228.260 88229.180 88230.180 88230.260
VWR®, TOC standard solutions
Standards Environmental Analysis Standards
Potassium hydrogen phthalate solutions.
  • Broad range of the most relevant standards from 5 to 2000 mg/L
  • Excellent quality
  • Ease of use
  • Reliable results
These TOC standards were developed for a broad and routine set of applications. They can be used on a wide variety of instruments including where instrument-specific standards are not required.
Total organic carbon (TOC) is the amount of carbon found in an organic compound. It is often used as a non specific indicator both for water quality and cleanliness of pharmaceutical manufacturing equipment.
TOC may also refer to the amount of organic carbon in soil, or in a geological formation.
TOC analysis acc. to EN 1484-H3.
This EU standard has been implemented for the analysis of TOC (replacing DIN 38409-H3). It regulates and provides guidelines for the determination of Total Organic Carbon (TOC) in different types of water (drinking, waste, ground, surface or lake). Our TOC standards (acc. to EN 1484) are especially designed for the easy calibration of TOC instruments according to this EU standard and to guarantee reliable analytical results.
Certifikace: Accredited according to ISO 17034, ISO 17025. ISO 9001 certified.
Upozornení: Store under normal laboratory conditions, at temperatures between 15 to 25 °C.