5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥98% (by TLC)

Supplier: Biotium

Synonyms: NHS-Fluorescein, FLUOS, 5(6)-FAM SE

90024
90024.EA 183 EUR
90024.
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥98% (by TLC)
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)

5(6)-FAM SE (full name: 5-(and-6)-Carboxyfluorescein, succinimidyl ester mixed isomers) is an amine-reactive green fluorescent dye widely used for labeling proteins or other molecules that contain a primary or secondary aliphatic amine. The coupling reaction is usually carried out at pH 8-9.5. The amide linkage from the coupling reaction is much more stable than the thiourea linkage formed from the coupling of an amine and an isothiocyanate.


  • λEx/λEm = 495/519 nm
  • Yellow solid soluble in DMSO or DMF
  • Store at -20°C and protect from light

Formula: C₂₅H₁₅NO₉
MW: 473.39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04119713
CAS Number: 117548-22-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb9033479a0382b801aee441231dac9140967bed&vid=eb9033479a0382b801aee441231dac9140967bed&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR