OpenLab CDS Software
Supplier: VWR Collection
906-0103EA 6510 EUR
906-0103 906-0104 906-0105 906-0106 906-0107 906-0108 906-0109 906-0110 906-0111 906-0112 906-0113 906-0114 906-0115 906-0119 SCSOK4626BA SCSOK4630BA
OpenLab CDS Software
Software Chromatography Software
Agilent OpenLab CDS is the next generation chromatography data system, designed for ease of use and productivity. OpenLab CDS offers the most comprehensive instrument control and data acquisition for Hitachi HPLC and amino acid analysers. In addition, it supports an expanded array of instrumentation from other vendors, for example Agilent LC, GC and LC/MS. Whether you like to control a single instrument or a huge network of different analytical instruments, OpenLab CDS grows with your needs.
  • Direct control of Hitachi HPLC and amino acid analyser