Dimethyl sulphoxide ≥99.8%, SPECTRONORM® for spectroscopy

Supplier: VWR Chemicals

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

Filtered through a 0.2 µm filter, packaged under nitrogen.

91258.260EA 127 EUR
91258.260 91258.320
Dimethyl sulphoxide ≥99.8%, SPECTRONORM® for spectroscopy
Dimethyl sulphoxide

SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
REACH: 01-2119431362-50
Merck Index: 13,03285

Order Now

Specification Test Results

Purity (GC) min. 99.8%
Evaporation residue max. 0.0004%
Water max. 0.05%
Color (Haz) max. 10
Acidity (meq/g) max. 0.0002
Alkalinity (meq/g) max. 0.0002
Transmission (270 nm) min. 35%
Transmission (280 nm) min. 50%
Transmission (310 nm) min. 80%
Transmission (330 nm) min. 90%
Transmission (from 350 nm) min. 97%
Absorbance (270 nm) max. 0.46
Absorbance (280 nm) max. 0.30
Absorbance (310 nm) max. 0.10
Absorbance (330 nm) max. 0.05
Absorbance (from 350 nm) max. 0.013


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR