Petroleum spirit, 30…60 °C, BAKER ANALYZED® for gas chromatography, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

9200.2500EA 317.26 EUR
9200.2500
Petroleum spirit, 30…60 °C, BAKER ANALYZED® for gas chromatography, J.T.Baker®
Petroleum spirit
Density: 0.7
Flash Pt: -10
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

Color (APHA) max. 10
Residue after Evaporation max. 5 ppm
Water (H2O) max. 0.05 %
Hydrocarbon oil index concentration (as RIVM oil reference standard) max. 0.5 mg/l
Total peaks between n-decane (C10H22) and n-tetracontane (C40H82) max. 0.5 mg/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR