Methanol, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®

Supplier: Avantor

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

9263.1000EA 122.4 EUR
9263.1000 9263.2500
Methanol, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Flash Pt: 11
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3 II
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) (corrected for water) min. 99.8 %
Acetone max. 0.001 %
Color (APHA) max. 10
Residue after Evaporation max. 1 ppm
Substances Darkened by H2SO4 passes test
Titrable Acid (meq/g) max. 0.0003
Titrable Base (meq/g) max. 0.0001
Water (H2O) max. 0.08 %
ECD Sensitive Impurities (as Heptachlor Epoxide)
Single Impurity Peak (pg/ml) max. 10
FID-Sensitive Impurities (as 2-Octanol)
Single Impurity Peak (ng/ml) max. 5

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR