VWR® IPASEPT® 70, Sterile
Supplier: VWR Chemicals
94006.1016EA 242 EUR
94006.1016
VWR® IPASEPT® 70, Sterile
Cleaning Agents Detergents and Surface Cleansers
Iso-propyl alcohol Ph. Eur. 70% V/V, with purified water Ph. Eur. 30% V/V, filtered at 0,2 μm.
  • Sterilisation by gamma irradiation
  • Sterility Assurance Level (SAL) 10⁻⁶
  • Filled under GMP conditions in cleanroom ISO Class 7
  • Certificate of Analysis available for each batch
  • Fine stream for precise applications, or wide spray for covering large areas
For surface spray cleaning, with or without wipes. Also for cleaning gloved hands before entry into cleanrooms. Ideal for wiping down entrances to controlled environments.