VWR® Catalase Reagent
Supplier: VWR Chemicals
941970ZAEA 33.8 EUR
941970ZA
VWR® Catalase Reagent
Microbiology Test Systems Enzymatic Tests
The catalase test can differentiate or identify microorganisms by checking their ability to produce catalase, an enzyme that protects microorganisms from toxic oxygen metabolites. Usually only aerobic microorganisms produce catalase.
  • A simple test to identify aerotolerant strains
  • Allows identification of Gram-positive cocci
  • Easy to use
  • Storage: 8 to 25 °C