Polymyxin B sulphate

Supplier: PanReac AppliChem

A0890.0001 A0890.0010
A0890.0001EA 62.2 EUR
A0890.0001 A0890.0010
Polymyxin B sulphate
Polymyxin B sulphate
Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
MDL Number: MFCD00131991
CAS Number: 1405-20-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR