Agar

Supplier: PanReac AppliChem

Synonyms: Agar Agar

A0917.1000 A0917.5000
A0917.1000EA 199 EUR
A0917.1000 A0917.5000
Agar
Agar
Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR