Methyl myristate 99

Supplier: Thermo Fisher Scientific

Synonyms: Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate

A10257.18 A10257.30
A10257.18EA 32.4 EUR
A10257.18 A10257.30
Methyl myristate 99
Methyl myristate

Purity: 99% CAS No.: 124-10-7 Molecular Formula: C15H30O2 Formula Weight: 242.40 Flash Point: 112°(234°F) Refractive Index: 1.4562 Melting Point: 18° Boiling Point: 323° MDL No.: MFCD00008983

Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 323 °C
Melting Pt: 18
Density: 0.855
Flash Pt: >112 °C (234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008983
CAS Number: 124-10-7
EINECS: 204-680-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR