8-Anilino-1-naphthalenesulphonic acid, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid

M-Clarity™ quality level = MQ200

A1028-100G A1028-25G A1028-5G
A1028-100GEA 842 EUR
A1028-100G A1028-25G A1028-5G
8-Anilino-1-naphthalenesulphonic acid, Sigma-Aldrich®
8-Anilino-1-naphthalenesulphonic acid
Formula: C₁₆H₁₃NO₃S
MW: 299.35 g/mol
Melting Pt: 215…217 °C (dec., lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00003998
CAS Number: 82-76-8
EINECS: 201-438-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01d5d119685a735fd50f704a22cc75f3876991d6&vid=3261625e787c3194013da235d669472298f459e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Conforms to Requirements
Gray to dark green or black
Appearance (Form) Powder
Solubility (Colour) Dark Green to Dark Brown to Black
Solubility (Turbidity)
50 mg/ml, DMF
Clear to Turbid
Proton NMR spectrum Conforms to Structure
Solvent Content ≤0.5%
Purity (HPLC) ≥97%
Recommended Retest Period 3 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR