Methyl-2,5-dihydroxybenzoate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Gentisic acid methyl ester, Methyl 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid methyl ester

A10407.06 A10407.14
A10407.06EA 92.3 EUR
A10407.06 A10407.14
Methyl-2,5-dihydroxybenzoate ≥98%
Methyl 2,5-dihydroxybenzoate

Purity: 98% CAS No.: 2150-46-1 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 86-87° MDL No.: MFCD00016464

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 86…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00016464
CAS Number: 2150-46-1
EINECS: 218-427-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR