L(+)-Cysteine ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

A10435.0B A10435.18 A10435.30 AB115193.0050
A10435.0BEA 605 EUR
A10435.0B A10435.18 A10435.30 ABCRAB115193.0050
L(+)-Cysteine ≥98%
L-Cysteine

Purity: 98+% CAS No.: 52-90-4 Molecular Formula: C3H7NO2S Formula Weight: 121.16 Physical Form: Crystalline powder Sensitivity: Air Sensitive Appearance: White Odour: Odourless Melting Point: ca 220° dec. Beilstein Reference: 1721408 Merck Reference: 14,2781 Optical Rotation: +7.9° (c=5 in 5N HCl) MDL No.: MFCD00064306

Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Boiling Pt: 254
Melting Pt: ∼ 220 °C (dec.)
Density: 1.523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR