4-Fluoro-3-nitrobenzoic acid 98

Supplier: Thermo Fisher Scientific
Warning

A10800.06 A10800.14 A10800.22
A10800.06EA 40.5 EUR
A10800.06 A10800.14 A10800.22
4-Fluoro-3-nitrobenzoic acid 98
4-Fluoro-3-nitrobenzoic acid

Purity: 98% CAS No.: 453-71-4 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Melting Point: 121-126° Boiling Point: 92°/15mm MDL No.: MFCD00007058

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Boiling Pt: 92 °C (15 mmHg)
Melting Pt: 121…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR