2,4,6-Trifluoroaniline ≥98%

Supplier: Thermo Fisher Scientific
Danger

A10964.03 A10964.06 A10964.14 A10964.22
A10964.03EA 25.8 EUR
A10964.03 A10964.06 A10964.14 A10964.22
2,4,6-Trifluoroaniline ≥98%
2,4,6-Trifluoroaniline

Purity: 98% CAS No.: 363-81-5 Molecular Formula: C6H4F3N Formula Weight: 147.10 Flash Point: 54°(129°F) Odour: Recognizable Boiling Point: 57°/22mm Beilstein Reference: 2831022 UN No.: 2926 MDL No.: MFCD00007650

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 57 °C (22 mmHg)
Melting Pt: 33…36 °C
Flash Pt: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007650
CAS Number: 363-81-5
EINECS: 206-660-8
UN: 2926
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR