Benzylamine 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: α-Aminotoluene

A10997.0B A10997.0I A10997.30 A10997.36
A10997.0BEA 126 EUR
A10997.0B A10997.0I A10997.30 A10997.36
Benzylamine 98
Benzylamine

Purity: 98+% CAS No.: 100-46-9 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 72°(162°F) Refractive Index: 1.5435 Melting Point: -43° Boiling Point: 184-185° Beilstein Reference: 741984 Merck Reference: 14,1125 UN No.: 2735 MDL No.: MFCD00008106

Formula: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Boiling Pt: 184…185 °C
Melting Pt: –43 °C
Density: 0.982
Flash Pt: 72 °C (162 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8 II
Merck Index: 13,01127

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR