Manganese(II) sulphate monohydrate ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate

A11809.0E A11809.36
A11809.0EEA 155 EUR
A11809.0E A11809.36
Manganese(II) sulphate monohydrate ≥97%
Manganese (II) sulphate monohydrate

Purity: 97% CAS No.: 10034-96-5 Molecular Formula: MnSO4.H2O Formula Weight: 169.02 (151.00anhy) Odour: Odourless pH: 3-3.5 Merck Reference: 14,5739 UN No.: 3077 MDL No.: MFCD00149159

Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Density: 2.95
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR